missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenylpyridine 95.0+%, TCI America™

Catalog No. m10945g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10945G Supplier TCI America Supplier No. M10945G
Only null left

Chemical Identifiers

CAS 10477-94-8
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number MFCD00191465
InChI Key VPFKZMSXKPTQJG-UHFFFAOYSA-N
PubChem CID 598136
IUPAC Name 3-methyl-5-phenylpyridine
SMILES CC1=CN=CC(=C1)C2=CC=CC=C2

Specifications

Boiling Point 160°C
Quantity 5 g
Formula Weight 169.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-5-phenylpyridine