CAS RN 10477-94-8
CAS RN 10477-94-8
IUPAC Name:
3-methyl-5-phenylpyridine
Synonyms:
3-methyl-5-phenylpyridine
Molecular Weight (g/mol):
169.23
Molecular Formula:
C12H11N
InChi Key:
VPFKZMSXKPTQJG-UHFFFAOYSA-N
SMILES:
CC1=CN=CC(=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Methyl-5-phenylpyridine 95.0+%, TCI America™
CAS: 10477-94-8 Molecular Formula: C12H11N Molecular Weight (g/mol): 169.227 MDL Number: MFCD00191465 InChI Key: VPFKZMSXKPTQJG-UHFFFAOYSA-N PubChem CID: 598136 IUPAC Name: 3-methyl-5-phenylpyridine SMILES: CC1=CN=CC(=C1)C2=CC=CC=C2