missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Quantity | 250 mg |
| Sensitivity | Air Sensitive |
| Formula Weight | 189.65 |
| Chemical Name or Material | 3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride |
Sécurité et manipulation
| Hazard Category | H315-H319 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only