missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-phenyl-1H-pyrazole, 97%

Catalog No. AAL0725003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0725003 1 g
AAL0725006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0725003 Supplier Thermo Scientific Chemicals Supplier No. L0725003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13788-84-6 | C10H10N2 | 158.20 g/mol

3-Methyl-4-phenyl-1H-pyrazole is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-phenyl-1H-pyrazole is used as intermediates.

Solubility
Soluble in DMSO, Methanol. Insoluble in water.

Notes
Protect from heat. Store away from air and oxidizing agents. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 13788-84-6
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00051752
InChI Key XTXZCNATVCIKTR-UHFFFAOYSA-N
Synonym 3-methyl-4-phenyl-1h-pyrazole, 3-methyl-4-phenylpyrazole, 3-methyl-4-phenylpyrazol, 3-methyl-4-phenyl-2h-pyrazole, acmc-1brbb, maybridge1_003313, 5-methyl-4-phenylpyrazole, pyrazole-based inhibitor 1, 3-methyl-4-phenyl-1h-pyrazol, 4-phenyl-5-methyl-1h-pyrazole
PubChem CID 139601
IUPAC Name 5-methyl-4-phenyl-1H-pyrazole
SMILES CC1=C(C=NN1)C1=CC=CC=C1

Specifications

Melting Point 140°C to 144°C
Quantity 1 g
Beilstein 3330
Solubility Information Soluble in DMSO,Methanol. Insoluble in water.
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material 3-Methyl-4-phenyl-1H-pyrazole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only