CAS RN 13788-84-6
IUPAC Name:
5-methyl-4-phenyl-1H-pyrazole
Synonyms:
3-methyl-4-phenyl-2H-pyrazole
Molecular Weight (g/mol):
158.2
Molecular Formula:
C10H10N2
InChi Key:
XTXZCNATVCIKTR-UHFFFAOYSA-N
SMILES:
CC1=C(C=NN1)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Methyl-4-phenyl-1H-pyrazole, 97%
CAS: 13788-84-6 Molecular Formula: C10H10N2 Molecular Weight (g/mol): 158.20 MDL Number: MFCD00051752 InChI Key: XTXZCNATVCIKTR-UHFFFAOYSA-N Synonym: 3-methyl-4-phenyl-1h-pyrazole,3-methyl-4-phenylpyrazole,3-methyl-4-phenylpyrazol,3-methyl-4-phenyl-2h-pyrazole,acmc-1brbb,maybridge1_003313,5-methyl-4-phenylpyrazole,pyrazole-based inhibitor 1,3-methyl-4-phenyl-1h-pyrazol,4-phenyl-5-methyl-1h-pyrazole PubChem CID: 139601 IUPAC Name: 5-methyl-4-phenyl-1H-pyrazole SMILES: CC1=C(C=NN1)C1=CC=CC=C1