missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitropyridine N-Oxide 97.0+%, TCI America™

Catalog No. p-7133860
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05275G 5 g
N052725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05275G Supplier TCI America Supplier No. N05275G
Only null left

Chemical Identifiers

CAS 1074-98-2
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00014626
InChI Key SSOURMYKACOBIV-UHFFFAOYSA-N
Synonym 3-methyl-4-nitropyridine n-oxide, 3-methyl-4-nitropyridine 1-oxide, 4-nitro-3-picoline n-oxide, 3-methyl-4-nitropyridine-n-oxide, 3-methyl-4-nitropyridin-1-ium-1-olate, 3-methyl-4-nitropyridine-1-oxide, 3-methyl-4-nitro-pyridine 1-oxide, 4-nitro-3-picoline 1-oxide, 4-nitro-3-picoline-n-oxide, pyridine, 3-methyl-4-nitro-, 1-oxide
PubChem CID 70633
IUPAC Name 3-methyl-4-nitro-1-oxidopyridin-1-ium
SMILES CC1=C(C=C[N+](=C1)[O-])[N+](=O)[O-]

Specifications

Melting Point 137°C
Quantity 5 g
Formula Weight 154.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-4-nitropyridine N-Oxide
RTECSNumber UT5775000
TSCA No