accessibility menu, dialog, popup

UserName

CAS RN 1074-98-2

CH3NONOO

CAS RN 1074-98-2

IUPAC Name: 3-methyl-4-nitropyridin-1-ium-1-olate
Synonyms: 3-methyl-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: SSOURMYKACOBIV-UHFFFAOYSA-N
SMILES: CC1=C[N+]([O-])=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results