missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7133500
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29151G 1 g
M29155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29151G Supplier TCI America Supplier No. M29151G
Only null left

Chemical Identifiers

CAS 80866-75-7
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007171
InChI Key KOVQGYQQVNCUBR-UHFFFAOYSA-N
Synonym 3-methyl-4-nitrobenzyl alcohol, 3-methyl-4-nitrophenyl methanol, 3-methyl-4-nitrobenzylalcohol, benzenemethanol, 3-methyl-4-nitro, acmc-20an0j, 4-nitro-3-methylbenzyl alcohol, 3-methyl-4-nitro-phenyl methanol, benzenemethanol,3-methyl-4-nitro, 3-methyl-4-nitro-phenyl-methanol, 3-methyl-4-nitrophenyl methanol #
PubChem CID 591360
IUPAC Name (3-methyl-4-nitrophenyl)methanol
SMILES CC1=C(C=CC(=C1)CO)[N+](=O)[O-]

Specifications

Melting Point 59°C
Quantity 1 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-4-nitrobenzyl Alcohol