missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-4-nitrobenzyl Alcohol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 59°C |
| Quantity | 5 g |
| Formula Weight | 167.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Methyl-4-nitrobenzyl Alcohol |
Safety and Handling
| TSCA | No |