missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-4-nitroaniline 97.0+%, TCI America™
$63.52 - $286.85
Chemical Identifiers
| CAS | 611-05-2 |
|---|---|
| Molecular Formula | C7H8N2O2 |
| Molecular Weight (g/mol) | 152.153 |
| InChI Key | XPAYEWBTLKOEDA-UHFFFAOYSA-N |
| Synonym | 4-nitro-m-toluidine, benzenamine, 3-methyl-4-nitro, 3-methyl-4-nitro-aniline, 3-methyl-4-nitro-phenylamine, 3-methyl-4-nitrophenylamine, 4-nitro-m-toluidin, 3-methyl4-nitroaniline, pubchem12456, 3-mehtyl-4-nitroanilin, 5-amino-2-nitrotoluene |
| PubChem CID | 11898 |
| IUPAC Name | 3-methyl-4-nitroaniline |
| SMILES | CC1=C(C=CC(=C1)N)[N+](=O)[O-] |
Chemical Identifiers
| 611-05-2 | |
| 152.153 | |
| 4-nitro-m-toluidine, benzenamine, 3-methyl-4-nitro, 3-methyl-4-nitro-aniline, 3-methyl-4-nitro-phenylamine, 3-methyl-4-nitrophenylamine, 4-nitro-m-toluidin, 3-methyl4-nitroaniline, pubchem12456, 3-mehtyl-4-nitroanilin, 5-amino-2-nitrotoluene | |
| 3-methyl-4-nitroaniline |
| C7H8N2O2 | |
| XPAYEWBTLKOEDA-UHFFFAOYSA-N | |
| 11898 | |
| CC1=C(C=CC(=C1)N)[N+](=O)[O-] |
Specifications
| 611-05-2 | |
| C7H8N2O2 | |
| 2660 | |
| XPAYEWBTLKOEDA-UHFFFAOYSA-N | |
| 3-methyl-4-nitroaniline | |
| 11898 | |
| ≥97.0% (GC) | |
| 3-Methyl-4-nitroaniline |
| 134°C | |
| 5 g | |
| 4-nitro-m-toluidine, benzenamine, 3-methyl-4-nitro, 3-methyl-4-nitro-aniline, 3-methyl-4-nitro-phenylamine, 3-methyl-4-nitrophenylamine, 4-nitro-m-toluidin, 3-methyl4-nitroaniline, pubchem12456, 3-mehtyl-4-nitroanilin, 5-amino-2-nitrotoluene | |
| CC1=C(C=CC(=C1)N)[N+](=O)[O-] | |
| 152.153 | |
| 152.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No