missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitroaniline, 95%

Catalog No. p-7046094
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1311206 5 g
AAL1311214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1311206 Supplier Thermo Scientific Chemicals Supplier No. L1311206
Only null left

CAS: 611-05-2 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-05-2
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00091833
InChI Key XPAYEWBTLKOEDA-UHFFFAOYSA-N
Synonym 4-nitro-m-toluidine, benzenamine, 3-methyl-4-nitro, 3-methyl-4-nitro-aniline, 3-methyl-4-nitro-phenylamine, 3-methyl-4-nitrophenylamine, 4-nitro-m-toluidin, 3-methyl4-nitroaniline, pubchem12456, 3-mehtyl-4-nitroanilin, 5-amino-2-nitrotoluene
PubChem CID 11898
IUPAC Name 3-methyl-4-nitroaniline
SMILES CC1=C(C=CC(=C1)N)[N+](=O)[O-]

Specifications

Melting Point 133°C to 137°C
Odor Characteristic
Quantity 5 g
UN Number UN2660
Beilstein 2209112
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 3-Methyl-4-nitroaniline
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)
EINECSNumber 210-247-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only