missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitro-1H-pyrazole, 97%

Catalog No. AAH3086006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3086006 5 g
AAH3086014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3086006 Supplier Thermo Scientific Chemicals Supplier No. H3086006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5334-39-4 | C4H5N3O2 | 127.10 g/mol

3-Methyl-4-nitropyrazole is a reactant used in the preparation of aminopyrazole derivatives as potent, selective and brain penetrant Leucine-rich repeat kinase 2(LRRK2) small molecule inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitropyrazole is a reactant used in the preparation of aminopyrazole derivatives as potent, selective and brain penetrant Leucine-rich repeat kinase 2(LRRK2) small molecule inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 5334-39-4
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00037864
InChI Key WTZYTQJELOHMMJ-UHFFFAOYSA-N
Synonym 3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitropyrazole, 1h-pyrazole, 3-methyl-4-nitro, 3-methyl-4-nitro pyrazole, 3-me-4-no2 pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j, 4-nitro-5-methyl-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole #
PubChem CID 79255
IUPAC Name 5-methyl-4-nitro-1H-pyrazole
SMILES CC1=C(C=NN1)[N+]([O-])=O

Specifications

Melting Point 131°C to 134°C
Boiling Point 325°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitro-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UQ7435000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only