accessibility menu, dialog, popup

UserName

CAS RN 5334-39-4

IUPAC Name: 5-methyl-4-nitro-1H-pyrazole
Synonyms: 3-ME-4-NO2 pyrazole
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: WTZYTQJELOHMMJ-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)