missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-4-heptanone 95.0+%, TCI America™
Supplier: TCI America M126710ML
Specifications
| 3-Methyl-4-heptanone | |
| Colorless | |
| C8H16O | |
| 10 mL | |
| sec-Butyl Propyl Ketone | |
| CCCC(=O)C(C)CC | |
| 128.22 | |
| 128.22 | |
| Liquid |
| 15726-15-5 | |
| 156°C | |
| MFCD00048721 | |
| 1224 | |
| NHIMSNHOEAVUKE-UHFFFAOYNA-N | |
| 3-methylheptan-4-one | |
| 27470 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 15726-15-5 | |
| 128.22 | |
| NHIMSNHOEAVUKE-UHFFFAOYNA-N | |
| 27470 | |
| CCCC(=O)C(C)CC |
| C8H16O | |
| MFCD00048721 | |
| sec-Butyl Propyl Ketone | |
| 3-methylheptan-4-one |
Safety and Handling
TSCA : No