accessibility menu, dialog, popup

UserName

CAS RN 15726-15-5

H3COCH3CH3

CAS RN 15726-15-5

IUPAC Name: 3-methylheptan-4-one
Synonyms: 4-heptanone, 3-methyl-
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: NHIMSNHOEAVUKE-UHFFFAOYNA-N
SMILES: CCCC(=O)C(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results