missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, ≥90.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012901
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012901 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012901 Supplier Merck Emd Millipore Supplier No. 167971ML
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 127-51-5
Density 0.93 g/mL (at 20°C)
Linear Formula C14H22 O
MDL Number MFCD00034582
Synonym alpha-Cetone; Isomethyl-alpha-ionone
Type Analytical Standard
Quantity 1 mL
Formula Weight 206.32
Percent Purity ≥90.0% (GC)
Grade Analytical Standard