missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-trimethylsiloxy-1-butyne, 97%

Catalog No. p-7059086
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4392209 10 g
AA4392218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4392209 Supplier Thermo Scientific Chemicals Supplier No. 04392209

CAS: 17869-77-1 | C8H16OSi | 156.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17869-77-1
Molecular Formula C8H16OSi
Molecular Weight (g/mol) 156.3
MDL Number MFCD00053867
InChI Key JNRUXZIXAXHXTN-UHFFFAOYSA-N
Synonym 3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne
PubChem CID 87344
IUPAC Name trimethyl(2-methylbut-3-yn-2-yloxy)silane
SMILES CC(C)(C#C)O[Si](C)(C)C

Specifications

Boiling Point 52°C (75 mmHg)
Odor Characteristic
Refractive Index 1.4052
Quantity 10 g
UN Number UN1993
Formula Weight 156.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-trimethylsiloxy-1-butyne
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 241-823-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only