accessibility menu, dialog, popup

UserName

CAS RN 17869-77-1

IUPAC Name: trimethyl[(2-methylbut-3-yn-2-yl)oxy]silane
Synonyms: trimethyl[(2-methylbut-3-yn-2-yl)oxy]silane
Molecular Weight (g/mol): 156.3
Molecular Formula: C8H16OSi
InChi Key: JNRUXZIXAXHXTN-UHFFFAOYSA-N
SMILES: CC(C)(O[Si](C)(C)C)C#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results