missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-pentanol 98.0+%, TCI America™

Supplier:  TCI America M0387100ML

 View more versions of this product

Catalog No. M0387100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methyl-3-pentanol
Yellow
C6H14O
100 mL
Diethylmethylcarbinol
CCC(C)(CC)O
102.177
102.18
Liquid
77-74-7
126°C
MFCD00004483
1987
FRDAATYAJDYRNW-UHFFFAOYSA-N
3-methylpentan-3-ol
6493
≥98.0% (GC)

Chemical Identifiers

77-74-7
102.177
FRDAATYAJDYRNW-UHFFFAOYSA-N
6493
CCC(C)(CC)O
C6H14O
MFCD00004483
Diethylmethylcarbinol
3-methylpentan-3-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : SA7450000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions