CAS RN 77-74-7
CAS RN 77-74-7
IUPAC Name:
3-methylpentan-3-ol
Synonyms:
3-methyl-3-pentanol
Molecular Weight (g/mol):
102.18
Molecular Formula:
C6H14O
InChi Key:
FRDAATYAJDYRNW-UHFFFAOYSA-N
SMILES:
CCC(C)(O)CC
1
–
1
of
1
results
1
3-Methyl-3-pentanol 98.0+%, TCI America™
CAS: 77-74-7 Molecular Formula: C6H14O Molecular Weight (g/mol): 102.177 MDL Number: MFCD00004483 InChI Key: FRDAATYAJDYRNW-UHFFFAOYSA-N Synonym: Diethylmethylcarbinol PubChem CID: 6493 IUPAC Name: 3-methylpentan-3-ol SMILES: CCC(C)(CC)O