missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-pentanol 98.0+%, TCI America™

Catalog No. M038725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M038725ML 25 mL
M0387100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M038725ML Supplier TCI America Supplier No. M038725ML

Chemical Identifiers

CAS 77-74-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004483
InChI Key FRDAATYAJDYRNW-UHFFFAOYSA-N
Synonym Diethylmethylcarbinol
PubChem CID 6493
IUPAC Name 3-methylpentan-3-ol
SMILES CCC(C)(CC)O

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 mL
UN Number 1987
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-pentanol
EINECSNumber (2)-0217
RTECSNumber SA7450000
TSCA Yes