missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-heptanol, TCI America™

Catalog No. M09555ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09555ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M09555ML Supplier TCI America Supplier No. M09555ML

Chemical Identifiers

CAS 5582-82-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00021841
InChI Key PQOSNJHBSNZITJ-UHFFFAOYSA-N
Synonym 3-methyl-3-heptanol, 3-heptanol, 3-methyl, 2-ethyl-2-hexanol, butylethylmethylcarbinol, 3-heptanol, 3-methyl-8ci 9ci, acmc-209lpn
PubChem CID 11710
IUPAC Name 3-methylheptan-3-ol
SMILES CCCCC(C)(CC)O

Specifications

Color Colorless
Boiling Point 64°C
Quantity 5 mL
UN Number 1987
Formula Weight 130.23
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-heptanol
EINECSNumber (2)-0217
TSCA No