accessibility menu, dialog, popup

UserName

CH3CH3HOH3C

CAS RN 5582-82-1

IUPAC Name: 3-methylheptan-3-ol
Synonyms: 3-methyl-3-heptanol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: PQOSNJHBSNZITJ-UHFFFAOYNA-N
SMILES: CCCCC(C)(O)CC
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)