missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-heptanol, 98%

Catalog No. AAB2095906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2095906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2095906 Supplier Thermo Scientific Chemicals Supplier No. B2095906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5582-82-1 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5582-82-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00021841
InChI Key PQOSNJHBSNZITJ-UHFFFAOYSA-N
Synonym 3-methyl-3-heptanol, 3-heptanol, 3-methyl, 2-ethyl-2-hexanol, butylethylmethylcarbinol, 3-heptanol, 3-methyl-8ci 9ci, acmc-209lpn
PubChem CID 11710
IUPAC Name 3-methylheptan-3-ol
SMILES CCCCC(C)(CC)O

Specifications

Density 0.83
Boiling Point 64°C (201 mmHg)
Flash Point 58°C (136°F)
Quantity 5 g
UN Number UN1987
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 3-Methyl-3-heptanol
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No

RUO – Research Use Only