missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pyridone 98.0+%, TCI America™

Catalog No. M14475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14475G 5 g
M144725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14475G Supplier TCI America Supplier No. M14475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1003-56-1 | C6H7NO | 109.128 g/mol

Chemical Identifiers

CAS 1003-56-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00039704
InChI Key MVKDNXIKAWKCCS-UHFFFAOYSA-N
Synonym 3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone
PubChem CID 70481
IUPAC Name 3-methyl-1H-pyridin-2-one
SMILES CC1=CC=CNC1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-2-pyridone