accessibility menu, dialog, popup

UserName

CH3HNO

CAS RN 1003-56-1

IUPAC Name: 3-methyl-1,2-dihydropyridin-2-one
Synonyms: 2(1H)-pyridinone, 3-methyl-
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: MVKDNXIKAWKCCS-UHFFFAOYSA-N
SMILES: CC1=CC=CNC1=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)