missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-phenylpyridine 98.0+%, TCI America™
$219.95 - $905.94
Chemical Identifiers
| CAS | 10273-90-2 |
|---|---|
| Molecular Formula | C12H11N |
| Molecular Weight (g/mol) | 169.23 |
| MDL Number | MFCD00006283 |
| InChI Key | BJATUPPYBZHEIO-UHFFFAOYSA-N |
| Synonym | 2-Phenyl-beta-picoline |
| PubChem CID | 66302 |
| IUPAC Name | 3-methyl-2-phenylpyridine |
| SMILES | CC1=CC=CN=C1C1=CC=CC=C1 |
Chemical Identifiers
| 10273-90-2 | |
| 169.23 | |
| BJATUPPYBZHEIO-UHFFFAOYSA-N | |
| 66302 | |
| CC1=CC=CN=C1C1=CC=CC=C1 |
| C12H11N | |
| MFCD00006283 | |
| 2-Phenyl-beta-picoline | |
| 3-methyl-2-phenylpyridine |
Specifications
| 10273-90-2 | |
| 160°C | |
| MFCD00006283 | |
| 2-Phenyl-beta-picoline | |
| CC1=CC=CN=C1C1=CC=CC=C1 | |
| 169.23 | |
| 169.23 | |
| Liquid |
| Yellow | |
| C12H11N | |
| 5 g | |
| BJATUPPYBZHEIO-UHFFFAOYSA-N | |
| 3-methyl-2-phenylpyridine | |
| 66302 | |
| ≥98.0% (GC) | |
| 3-Methyl-2-phenylpyridine |
Safety and Handling
TSCA : No