accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 10273-90-2

CH3N

CAS RN 10273-90-2

IUPAC Name: 3-methyl-2-phenylpyridine
Synonyms: 3-methyl-2-phenylpyridine
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: BJATUPPYBZHEIO-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1C1=CC=CC=C1
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.