missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-phenylpyridine 98.0+%, TCI America™

Catalog No. M09325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09325G 5 g
M093225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09325G Supplier TCI America Supplier No. M09325G

Chemical Identifiers

CAS 10273-90-2
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006283
InChI Key BJATUPPYBZHEIO-UHFFFAOYSA-N
Synonym 2-Phenyl-beta-picoline
PubChem CID 66302
IUPAC Name 3-methyl-2-phenylpyridine
SMILES CC1=CC=CN=C1C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 160°C
Quantity 5 g
Formula Weight 169.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-phenylpyridine