missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-oxazolidone 99.0+%, TCI America™
$130.48 - $1222.83
Chemical Identifiers
| CAS | 19836-78-3 |
|---|---|
| Molecular Formula | C4H7NO2 |
| Molecular Weight (g/mol) | 101.105 |
| MDL Number | MFCD00009759 |
| InChI Key | VWIIJDNADIEEDB-UHFFFAOYSA-N |
| Synonym | 2-Oxo-3-methyloxazolidine |
| PubChem CID | 29831 |
| ChEBI | CHEBI:39997 |
| IUPAC Name | 3-methyl-1,3-oxazolidin-2-one |
| SMILES | CN1CCOC1=O |
Chemical Identifiers
| 19836-78-3 | |
| 101.105 | |
| VWIIJDNADIEEDB-UHFFFAOYSA-N | |
| 29831 | |
| 3-methyl-1,3-oxazolidin-2-one |
| C4H7NO2 | |
| MFCD00009759 | |
| 2-Oxo-3-methyloxazolidine | |
| CHEBI:39997 | |
| CN1CCOC1=O |
Specifications
| 19836-78-3 | |
| Yellow | |
| MFCD00009759 | |
| VWIIJDNADIEEDB-UHFFFAOYSA-N | |
| 3-methyl-1,3-oxazolidin-2-one | |
| 101.105 | |
| CHEBI:39997 | |
| ≥99.0% (GC) | |
| 3-Methyl-2-oxazolidone |
| 88°C | |
| C4H7NO2 | |
| 2-Oxo-3-methyloxazolidine | |
| CN1CCOC1=O | |
| 25 g | |
| 29831 | |
| 101.11 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-5494
RTECSNumber : RQ3550000
TSCA : Yes