missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-oxazolidone 99.0+%, TCI America™

Supplier:  TCI America M0781100G

 View more versions of this product

Catalog No. M0781100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methyl-2-oxazolidone
88°C
Yellow
MFCD00009759
VWIIJDNADIEEDB-UHFFFAOYSA-N
3-methyl-1,3-oxazolidin-2-one
101.105
CHEBI:39997
≥99.0% (GC)
19836-78-3
10.6 (50% H2O soln.)
C4H7NO2
2-Oxo-3-methyloxazolidine
CN1CCOC1=O
100 g
29831
101.11
Liquid

Chemical Identifiers

19836-78-3
101.105
VWIIJDNADIEEDB-UHFFFAOYSA-N
29831
3-methyl-1,3-oxazolidin-2-one
C4H7NO2
MFCD00009759
2-Oxo-3-methyloxazolidine
CHEBI:39997
CN1CCOC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5494

RTECSNumber : RQ3550000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions