missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-hexanone 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 145°C |
| Quantity | 10 mL |
| UN Number | 1224 |
| Formula Weight | 114.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Methyl-2-hexanone |
Safety and Handling
| EINECSNumber | (2)-0542 |
| TSCA | No |