accessibility menu, dialog, popup

UserName

CAS RN 2550-21-2

H3CCH3CH3O

CAS RN 2550-21-2

IUPAC Name: 3-methylhexan-2-one
Synonyms: 1-methylbutyl methyl ketone
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: GYWYASONLSQZBB-UHFFFAOYNA-N
SMILES: CCCC(C)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results