missing translation for 'onlineSavingsMsg'
Learn More

Methyl Salicylate, NF, 98-102%, Spectrum™ Chemical

Catalog No. p-7157814
Change view
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604454 500 mL Steel Can
18604455 4 L Steel Can
18604457 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604454 Supplier Spectrum Chemical Mfg Cor Supplier No. ME170500MLGL

ME170, 119-36-8, C8H8O3

Spectrum™ Chemical Methyl Salicylate, NF is used to treat minor aches and pains of the muscles and joints. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

CAS Max % 100%
Packaging Steel Can
Quantity 500 mL
Percent Purity 98 to 102%
Grade NF
Physical Form Liquid