missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-furoic acid, 98+%

Catalog No. AAL0176306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0176306 5 g
AAL0176303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0176306 Supplier Thermo Scientific Chemicals Supplier No. L0176306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4412-96-8 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4412-96-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00014111
InChI Key BNYIQEFWGSXIKQ-UHFFFAOYSA-N
Synonym 3-methyl-2-furoic acid, 2-furancarboxylic acid, 3-methyl, 3-methyl-2-furancarboxylic acid, unii-5jp68392hm, 3-methyl-furan-2-carboxylic acid, 3-methyl-2-furoicacid, methylpyromucic acid, 3-methyl furoic acid, 3-methylpyromucic acid, acmc-20a5im
PubChem CID 78127
IUPAC Name 3-methylfuran-2-carboxylic acid
SMILES CC1=C(OC=C1)C(O)=O

Specifications

Melting Point 133°C to 137°C
Quantity 5 g
Beilstein 116183
Formula Weight 126.11
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-furoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-571-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only