accessibility menu, dialog, popup

UserName

CAS RN 4412-96-8

IUPAC Name: 3-methylfuran-2-carboxylic acid
Synonyms: 3-methylfuran-2-carboxylic acid
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: BNYIQEFWGSXIKQ-UHFFFAOYSA-N
SMILES: CC1=C(OC=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)