missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-furancarboxylic Acid 98.0+%, TCI America™

Catalog No. M23761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23761G 1 g
M23765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23761G Supplier TCI America Supplier No. M23761G

Chemical Identifiers

CAS 4412-96-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00014111
InChI Key BNYIQEFWGSXIKQ-UHFFFAOYSA-N
Synonym 3-methyl-2-furoic acid, 2-furancarboxylic acid, 3-methyl, 3-methyl-2-furancarboxylic acid, unii-5jp68392hm, 3-methyl-furan-2-carboxylic acid, 3-methyl-2-furoicacid, methylpyromucic acid, 3-methyl furoic acid, 3-methylpyromucic acid, acmc-20a5im
PubChem CID 78127
IUPAC Name 3-methylfuran-2-carboxylic acid
SMILES CC1=C(OC=C1)C(O)=O

Specifications

Melting Point 136°C
Color White
Quantity 1 g
Formula Weight 126.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-2-furancarboxylic Acid