missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butenal 97.0+%, TCI America™

Catalog No. p-7132352
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07705ML 5 mL
M077025ML 25 mL
M0770100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M07705ML Supplier TCI America Supplier No. M07705ML
Only null left

Chemical Identifiers

CAS 107-86-8
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00010291
InChI Key SEPQTYODOKLVSB-UHFFFAOYSA-N
Synonym 3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
PubChem CID 61020
ChEBI CHEBI:15825
IUPAC Name 3-methylbut-2-enal
SMILES CC(=CC=O)C

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 135°C
Quantity 5 mL
UN Number 1989
Formula Weight 84.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butenal
EINECSNumber (2)-2461
TSCA Yes
Recommended Storage Refrigerator