accessibility menu, dialog, popup

UserName

CH3CH3O

CAS RN 107-86-8

IUPAC Name: 3-methylbut-2-enal
Synonyms: prenal senecioaldehyde 2-isopentenal senecialdehyde
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: SEPQTYODOKLVSB-UHFFFAOYSA-N
SMILES: CC(C)=CC=O
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (5)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (6)