missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Methyl-2-butanol, 98%

Catalog No. p-7044228
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0534722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0534722 Supplier Thermo Scientific Chemicals Supplier No. L0534722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 598-75-4 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
3-Methyl-2-butanol has been used as a reference for microbially produced, volatile organic compounds (MVOC) in GC-MS analyses. It acts as a plant metabolite and polar solvent.

Solubility
Miscible in Ether, Alcohols. Slightly soluble in water. (2.8g/100mL, 30°C)

Notes
Incompatible materials are Strong oxidizing agents, Reducing agents.

Chemical Identifiers

CAS 598-75-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004527
InChI Key MXLMTQWGSQIYOW-UHFFFAOYNA-N
Synonym 3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol
PubChem CID 11732
ChEBI CHEBI:77517
IUPAC Name 3-methylbutan-2-ol
SMILES CC(C)C(C)O

Specifications

Density 0.818
Boiling Point 112°C
Flash Point 26°C (78°F)
Refractive Index 1.409
Quantity 100 g
UN Number UN1105
Beilstein 1718800
Merck Index 14,6031
Solubility Information Miscible in Ether,Alcohols. Slightly soluble in water. (2.8g/100mL,30°C)
Formula Weight 88.15
Percent Purity 98%
Chemical Name or Material (+/-)-3-Methyl-2-butanol
Show More Show Less
Hazard Category H226-H332
Hazard Statement GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
EINECSNumber 209-950-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only