accessibility menu, dialog, popup

UserName

CAS RN 598-75-4

IUPAC Name: 3-methylbutan-2-ol
Synonyms: 3-methyl-2-butanol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: MXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILES: CC(C)C(C)O
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)