missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%

Catalog No. p-7037347
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5606106 5 g
AAH5606118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5606106 Supplier Thermo Scientific Chemicals Supplier No. H5606106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38894-11-0 | C8H12ClN3OS | 233.714 g/mol

It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 38894-11-0
Molecular Formula C8H12ClN3OS
Molecular Weight (g/mol) 233.714
MDL Number MFCD00149370
InChI Key IYXXQOGEFHAQGU-PIHABLKOSA-N
Synonym 2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 3-methyl-2-benzothiazolinone hydrazone hcl monohydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, e-3-methyl-1,3-benzothiazol-2-ylidene hydrazine hydrate hydrochloride, e-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 2z-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrate hydrochloride
PubChem CID 6508671
IUPAC Name (E)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrate;hydrochloride
SMILES CN1C2=CC=CC=C2SC1=NN.O.Cl

Specifications

Melting Point 270°C to 274°C
Quantity 5 g
UN Number UN2811
Beilstein 4722243
Solubility Information Soluble in water.
Formula Weight 233.72 (215.70 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 238-428-7
RTECSNumber DL7160200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only