missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%

Catalog No. AC414020100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414020100 10 g Glass bottle
AC414020500 50 g Glass bottle
AC414022500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC414020100 Supplier Thermo Scientific Chemicals Supplier No. 414020100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38894-11-0 | HCl·H2O | 233.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38894-11-0
Molecular Formula HCl·H2O
Molecular Weight (g/mol) 233.72
MDL Number MFCD00149370
InChI Key IYXXQOGEFHAQGU-PIHABLKOSA-N
Synonym 2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 3-methyl-2-benzothiazolinone hydrazone hcl monohydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, e-3-methyl-1,3-benzothiazol-2-ylidene hydrazine hydrate hydrochloride, e-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 2z-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrate hydrochloride
PubChem CID 6508671
IUPAC Name (E)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrate;hydrochloride
SMILES CN1C2=CC=CC=C2SC1=NN.O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 270.0°C to 276.0°C
Color White
Loss on Drying 8% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Sensitivity (to hydroxyl groups) Passes Test
Solubility Information Solubility in water: soluble
Formula Weight 233.72
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.

RUO – Research Use Only