missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1H-pyrazole-4-carboxaldehyde, 97%

CAS: 112758-40-4 | C5H6N2O | 110.12 g/mol

Supplier:  Thermo Scientific Chemicals H3254703

 View more versions of this product

Catalog No. AAH3254703


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methyl-1H-pyrazole-4-carboxaldehyde
C5H6N2O
1 g
NWDMGTFNIOCVDU-UHFFFAOYSA-N
5-methyl-1H-pyrazole-4-carbaldehyde
2760056
97%
112758-40-4
MFCD01313822
3-methyl-1h-pyrazole-4-carbaldehyde, 3-methyl-1h-pyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-methyl, 3-methyl-2h-pyrazole-4-carbaldehyde, 5-methylpyrazole-4-carbaldehyde, pubchem16065, acmc-1brby, 4-formyl-3-methylpyrazole
CC1=C(C=O)C=NN1
110.12
110.12

Chemical Identifiers

112758-40-4
110.12
NWDMGTFNIOCVDU-UHFFFAOYSA-N
2760056
CC1=C(C=O)C=NN1
C5H6N2O
MFCD01313822
3-methyl-1h-pyrazole-4-carbaldehyde, 3-methyl-1h-pyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-methyl, 3-methyl-2h-pyrazole-4-carbaldehyde, 5-methylpyrazole-4-carbaldehyde, pubchem16065, acmc-1brby, 4-formyl-3-methylpyrazole
5-methyl-1H-pyrazole-4-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only