missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1H-pyrazole-4-carboxaldehyde, 97%

Catalog No. p-7031462
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32547MD 250 mg
AAH3254703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32547MD Supplier Thermo Scientific Chemicals Supplier No. H32547MD
Only null left

CAS: 112758-40-4 | C5H6N2O | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112758-40-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01313822
InChI Key NWDMGTFNIOCVDU-UHFFFAOYSA-N
Synonym 3-methyl-1h-pyrazole-4-carbaldehyde, 3-methyl-1h-pyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carboxaldehyde, 3-methylpyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde, 3-methyl, 3-methyl-2h-pyrazole-4-carbaldehyde, 5-methylpyrazole-4-carbaldehyde, pubchem16065, acmc-1brby, 4-formyl-3-methylpyrazole
PubChem CID 2760056
IUPAC Name 5-methyl-1H-pyrazole-4-carbaldehyde
SMILES CC1=C(C=O)C=NN1

Specifications

Quantity 250 mg
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 3-Methyl-1H-pyrazole-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only