missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-5-pyrazolone 98.0+%, TCI America™

Catalog No. M068725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M068725G 25 g
M0687500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M068725G Supplier TCI America Supplier No. M068725G

Chemical Identifiers

CAS 89-25-8
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00003138
InChI Key QELUYTUMUWHWMC-UHFFFAOYSA-N
Synonym edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
PubChem CID 4021
ChEBI CHEBI:31530
IUPAC Name 5-methyl-2-phenyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1)C2=CC=CC=C2

Specifications

Melting Point 129°C
Color White-Yellow
Boiling Point 287°C
Quantity 25 g
Formula Weight 174.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-1-phenyl-5-pyrazolone
EINECSNumber (5)-0287
RTECSNumber UQ9625000
TSCA Yes