missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-3-pentanol 98.0+%, TCI America™

Catalog No. p-7132965
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20115G 5 g
M201125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20115G Supplier TCI America Supplier No. M20115G

Chemical Identifiers

CAS 10415-87-9
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00021825
InChI Key AEJRTNBCFUOSEM-UHFFFAOYSA-N
PubChem CID 61516
IUPAC Name 3-methyl-1-phenylpentan-3-ol
SMILES CCC(C)(CCC1=CC=CC=C1)O

Specifications

Color Colorless
Boiling Point 130°C
Quantity 5 g
Formula Weight 178.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-phenyl-3-pentanol
EINECSNumber (3)-1044
RTECSNumber SA7624000
TSCA Yes