accessibility menu, dialog, popup

UserName

CAS RN 10415-87-9

IUPAC Name: 3-methyl-1-phenylpentan-3-ol
Synonyms: phenethyl methyl ethyl carbinol
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: AEJRTNBCFUOSEM-UHFFFAOYNA-N
SMILES: CCC(C)(O)CCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results