missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-butanone 98.0+%, TCI America™

Catalog No. p-7118814
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10295ML 5 mL
B102925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10295ML Supplier TCI America Supplier No. B10295ML

Chemical Identifiers

CAS 2893-05-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00040487
InChI Key ZATIMOAGKJEVGN-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-2-butanone, benzyl isopropyl ketone, isopropylbenzyl ketone, 2-butanone, 3-methyl-1-phenyl, 1-phenyl-3-methyl-2-butanone, unii-81804s3mkm, isopropyl benzyl ketone, isopropylbenzylketon, 5-nitrophenyl laurate, 2-methyl-4-phenyl-3-butanone
PubChem CID 76163
IUPAC Name 3-methyl-1-phenylbutan-2-one
SMILES CC(C)C(=O)CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 mL
Formula Weight 162.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-phenyl-2-butanone