accessibility menu, dialog, popup

UserName

CAS RN 2893-05-2

IUPAC Name: 3-methyl-1-phenylbutan-2-one
Synonyms: 3-methyl-1-phenylbutan-2-one
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: ZATIMOAGKJEVGN-UHFFFAOYSA-N
SMILES: CC(C)C(=O)CC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)